Learn More
Testo™ Temperature Probe with Aluminum Sleeve
Suitable for air temperature measurements and also for temperature measurements in liquids and pastes
Brand: Testo™ 0628 7503
This item is not returnable.
View return policy
Description
- Because of the NTC sensor element, the probe provides precise temperature measurements with an accuracy of up to ±0.2 °C/F
- Features a 2.4 m cable
- With this probe you are ideally equipped for measuring temperatures which are not in close proximity to the Testo Saveris 2-T2 Wi-Fi data logger
- The temperature probe is rated protection class IP 65 (waterproof)
- Measuring range: -22 to 194 °F (-30 to +90 °C)
- For use in laboratories and food industries
Specifications
IP 65 | |
190 sec. | |
2.4 m (Cable), 40.64 mm (Probe Shaft) |
±0.2°C (0 to +70 °C), ±0.5 °C (Remaining Range) | |
-30°C to +90°C |