Learn More
Testo™ Temperature Probe with Aluminum Sleeve
Suitable for air temperature measurements and also for temperature measurements in liquids and pastes
Brand:  Testo™ 0628 7503
Product Code. 11341174
  • 535.00 SEK / Each
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Because of the NTC sensor element, the probe provides precise temperature measurements with an accuracy of up to ±0.2 °C/F
  • Features a 2.4 m cable
  • With this probe you are ideally equipped for measuring temperatures which are not in close proximity to the Testo Saveris 2-T2 Wi-Fi data logger
  • The temperature probe is rated protection class IP 65 (waterproof)
  • Measuring range: -22 to 194 °F (-30 to +90 °C)
  • For use in laboratories and food industries
Specifications
Specifications
IP 65
190 sec.
2.4 m (Cable), 40.64 mm (Probe Shaft)
±0.2°C (0 to +70 °C), ±0.5 °C (Remaining Range)
-30°C to +90°C