Learn More
Memmert™ Temperature Restriction for UF110plus
Optional temperature restriction between 60°C and 250°C.
Brand:  Memmert™ UF110plusA8
Product Code. 17558935
  • 1900.00 SEK / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • 1 Choice
  • Temperatures: 60°C, 70°C, 80°C, 95°C, 100°C, 120°C, 160°C, 180°C, 200°C, or 250°C
Specifications
Specifications
UF110plus Oven
Temperature Restriction
Documents
One moment while we fetch your results.
Product Title
Memmert™ Temperature Restriction for UF110plus > For Use With: UF110plus