Learn More
Cole-Parmer™ Tensiometer, Capillary Surface Tension Apparatus
Quickly and easily determine surface tension based on height of liquid in capillary tube
Brand:  Cole-Parmer™ 59780-90
Additional Details : Weight : 0.27000kg
Product Code. 15248438
  • 4970.00 SEK / Each
Estimated Shipment: 28-08-2024
to see stock.
Add to basket
Description
Description
  • Clear, easy-to-read borosilicate glass capillary tube
  • Included rubber stopper centers glass capillary tube in the outer cylinder
  • Apparatus is comprised of an engraved 250-mm borosilicate glass capillary tube (graduated from 0 to 10 cm in 1-mm divisions) and an outer tube.
  • The capillary tube is positioned in the center of the outer tube by means of a rubber stopper.
  • The outer tube accommodates a pipette bulb to expel air.
Specifications
Specifications
Tensiometer, Capillary Surface Tension Apparatus
250 mm borosilicate glass capillary tube, rubber stopper, and outer cylinder