Learn More
Terumo™ Surflo™ Winged Infusion Sets
Secure, safe and simple I.V. therapy permits continuous access to a vein with minimum discomfort
9045.00 SEK
Product Code | Brand | Color | Needle Gauge | Product Type | Length (English) Tube | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Needle Gauge | Product Type | Length (English) Tube | Price | Quantity & Availability | ||
16398477
|
Terumo™
1SV27EL |
Gray | 27 G | Regular Wall needle | 8 in. |
9045.00 SEK
Pack of 1000 |
Estimated Shipment: 28-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
- Siliconized needle bonded to flexible wings
- Soft tubing attached to a Luer fitting with threaded venting cap
- Needles bonded to wings at slight downward angle—better conform to body contours and reduce trauma to tunica intima
- Color-coded wings according to needle gauge and embossed with gauge size
- Soft polyvinyl chloride tubing resists kinking
Order Info
22-254-038 and 22-254-039 have a latex cap covering the end of the Luer fitting. With these sets, solutions can be intermittently injected (with a hypodermic syringe) directly through the cap and into the tubing.
Specifications
PVC | |
Sterile |