Learn More
Terumo™ Surflo™ Winged Infusion Sets
Secure, safe and simple I.V. therapy permits continuous access to a vein with minimum discomfort
9045.00 SEK
Specifications
Material PVC
Sterility Sterile
View More Specs
Products 1
Product Code Brand Color Needle Gauge Product Type Length (English) Tube Price Quantity & Availability  
Product Code Brand Color Needle Gauge Product Type Length (English) Tube Price Quantity & Availability  
16398477
View Documents
Terumo™
1SV27EL
Gray 27 G Regular Wall needle 8 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Siliconized needle bonded to flexible wings
  • Soft tubing attached to a Luer fitting with threaded venting cap
  • Needles bonded to wings at slight downward angle—better conform to body contours and reduce trauma to tunica intima
  • Color-coded wings according to needle gauge and embossed with gauge size
  • Soft polyvinyl chloride tubing resists kinking
Order Info
22-254-038 and 22-254-039 have a latex cap covering the end of the Luer fitting. With these sets, solutions can be intermittently injected (with a hypodermic syringe) directly through the cap and into the tubing.
Specifications
Specifications
PVC