Learn More
Elma™ Test Tube Holders
Ensures that tubes can be securely positioned and arranged in the Elma Basket.
Brand: Elma™ 1112123
Additional Details : Weight : 0.76200kg
Description
- Diverse Elma holders ensure that tubes can be securely positioned and arranged in the Elma Basket
- Optimum positioning of the items inside the bath is also a prerequisite for achieving the best possible cleaning results
- Allows items to be individually positioned
- Lets you clearly structure your cleaning process
- Prevents scratches on the tank floor
- Holders are available for various applications
- Special solutions are available (e.g. for conical flasks)
Specifications
Test Tube Holder | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY | |
Stainless Steel | |
150/300 |