Learn More
Elma™ Test Tube Holders
Ensures that tubes can be securely positioned and arranged in the Elma Basket.
769.19 SEK - 1185.00 SEK
Specifications
Type | Test Tube Holder |
---|---|
For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
Material | Stainless Steel |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
17789899
|
Elma™
1111973 |
30/80 |
769.19 SEK
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
17851297
|
Elma™
1112084 |
100/120 |
1020.00 SEK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
17801357
|
Elma™
1112123 |
150/300 |
1185.00 SEK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
Description
- Diverse Elma holders ensure that tubes can be securely positioned and arranged in the Elma Basket
- Optimum positioning of the items inside the bath is also a prerequisite for achieving the best possible cleaning results
- Allows items to be individually positioned
- Lets you clearly structure your cleaning process
- Prevents scratches on the tank floor
- Holders are available for various applications
- Special solutions are available (e.g. for conical flasks)
Specifications
Test Tube Holder | |
Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY | |
Stainless Steel |