missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, BHT Stabilized, Reagent Grade, ACS, Ph Eur, Scharlab

Product Code. 11302575
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Complies with American Chemical Society and European Pharmacopeia specifications. Tetrahydrofuran, BHT Stabilized, Reagent Grade, ACS, Ph Eur, Scharlab is stabilized with 250 ppm of 2,6-Di-tert-butyl-4-methylphenol and is packed in a glass bottle.

Reagent grade chemicals are the most frequently used grade for laboratory analytical work. They correspond to a grade of high quality chemicals for laboratory and specialized industrial use.

Specifications

Melting Point -108.5°C
CAS 109-99-9
Color Colorless
Acidity Max. 0.0003meq/g
Alkalinity Max. 0.0002meq/g
Appearance Clear
Density 0.89g/cm3
Boiling Point 65°C to 66°C
Quantity 1 L
Flash Point −21.5°C
Identification Passes test
Ignition Point 215°C
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.025%
Molecular Formula C4H8O
MDL Number MFCD00005356
Vapor Pressure 173 hPa
UN Number 2056
Water Max. 0.03%
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
Solubility Information Miscible in water
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.11
PubChem CID 8028
ChEBI CHEBI:26911
Formula Weight 72.11g/mol
Percent Purity Min. 99.8%
Grade Reagent
Chemical Name or Material Tetrahydrofuran
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'