missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TEX55 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP2-37991PEP
1968.00 SEK valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human C3ORF30. The TEX55 Recombinant Protein Antigen is derived from E. coli. The TEX55 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.This is a blocking peptide for NBP2-37991. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

Specifications
152405 | |
Chromatography | |
0.5mg/mL | |
PBS and 1M Urea, pH 7.4. | |
C3ORF30 | |
27kDa | |
0.1 mL | |
E.Coli | |
QAERRTSGQIDGRLAMPSDQRGSRQTDHRMAGQSERRASEQMDRRMSGEAERRTSEQITHRLSKLSERRPSVQIDSGSSVPSDQSP |
Human | |
>80% | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
Unlabeled | |
C3orf30 | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-37991. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction