Learn More
Biochrom Thermal Printer Paper
Thermal Printer Paper for use with Ultrospec 2100, 3100 and 6300 Pro.
Marque: Biochrom 80200980
Informations supplémentaires : Weight : 0.38000kg
Description
- Thermal printer paper
- For use with Ultrospec 2100, 3100 and 6300 Pro
- 5 rolls per pack
Spécification
Paper | |
White | |
Not Autoclavable | |
Ultrospec 2100, 3100 and 6300 Pro | |
Thermal Printer Paper | |
Non-sterile |
No | |
White | |
Single-use | |
Roll | |
5 Pk. |