Learn More
GFL Gesellschaft Fuer Labortec™ Thermolab™ 10770 Shaking Waterbath
Brand: GFL Gesellschaft Fuer Labortec™ 1070
This item is not returnable.
View return policy
Specifications
Reciprocating | |
Shaking Water Bath | |
1.3 kW | |
625 mm | |
270 mm | |
556 mm | |
28 / 36 kg | |
230 V |
5°C to 100°C | |
2 to 50 rpm | |
2 to 50rpm | |
175 mm | |
100 mm | |
175 mm | |
230 V | |
50/60 Hz |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GFL Gesellschaft Fuer Labortec™ Thermolab™ 10770 Shaking Waterbath > Weight: 28 / 36kg
Spot an opportunity for improvement?Share a Content Correction