missing translation for 'onlineSavingsMsg'
Learn More

Thiamet G, MedChemExpress

MedChemExpress Thiamet G is a potent and selective inhibitor of O-GlcNAcase (OGA), which acts to remove O-GlcNAc from modified proteins, with Ki of 20 nM for human OGA.

Encompass_Preferred

1275.97 SEK - 8480.52 SEK

Specifications

Chemical Name or Material Thiamet G
Physical Form Powder
CAS 1009816-48-1
Color White
Purity Grade Notes Research
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16473316
Encompass_Preferred
View Documents
Medchem Express
HY-12588
25 mg
4668.18 SEK
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16427977
Encompass_Preferred
View Documents
Medchem Express
HY-12588
5 mg
1275.97 SEK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16421467
Encompass_Preferred
View Documents
Medchem Express
HY-12588
50 mg
8480.52 SEK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16415258
Encompass_Preferred
View Documents
Medchem Express
HY-12588
10 mg
2116.25 SEK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thiamet G is a potent and selective inhibitor of O-GlcNAcase (OGA), which acts to remove O-GlcNAc from modified proteins, with Ki of 20 nM for human OGA.
Specifications

Specifications

Thiamet G
1009816-48-1
Research
Cancer-Kinase/protease
O[C@H]1[C@H](O)[C@@]2([H])N=C(NCC)S[C@@]2([H])O[C@@H]1CO
248.3
Research
Powder
White
C9H16N2O4S
H2O : ≥ 50 mg/mL (201.37 mM) ∣DMSO : ≥ 45 mg/mL (181.23 mM)
248.3
98.32%
Videos
Safety and Handling

Safety and Handling

H317

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Thiamet G, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.