missing translation for 'onlineSavingsMsg'
Learn More

Thiolactic Acid, ≥95%, MP Biomedicals™

Product Code. 11476131
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 79-42-5
Color Colorless
Assay Percent Range ≥95%
Molecular Formula C3H6O2S
Quantity 100 mL
Synonym 2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
InChI Key PMNLUUOXGOOLSP-UHFFFAOYNA-N
SMILES CC(S)C(O)=O
IUPAC Name 2-sulfanylpropanoic acid
Molecular Weight (g/mol) 106.14
PubChem CID 62326
ChEBI CHEBI:47872
Formula Weight 106.14
Physical Form Liquid
Chemical Name or Material Thiolactic Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.