Learn More
Lenz Laborglasintrumente™ Three-Neck Round-Bottom Flasks
Three-neck round-bottom reaction flasks with two parallel side necks
Brand: Lenz Laborglasintrumente™ 03305476
This item is not returnable.
View return policy
Description
- Reaction flasks acc. to DIN 12392 and in addition to DIN
- With two parallel side necks
- With conical ground joints
- With inscription label
- Material: DURAN™
Specifications
Center neck- NS 45/40 , Side necks -NS 29/32 | |
Borosilicate Glass (3.3) | |
Reaction flasks |
3000 mL | |
1 | |
185 mm |