Learn More
Lenz Laborglasintrumente™ Three-Neck Round-Bottom Flasks
Three-neck round-bottom reaction flasks with two parallel side necks
Brand:  Lenz Laborglasintrumente™ 03304479
 View more versions of this product
Product Code. 10718841
  • 2705.00 SEK / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Description
  • Reaction flasks acc. to DIN 12392 and in addition to DIN
  • With two parallel side necks
  • With conical ground joints
  • With inscription label
  • Material: DURAN™
Specifications
Specifications
Center neck- NS 29/32, Side necks - NS 29/32
Borosilicate Glass (3.3)
Reaction flasks
4000 mL
1
207 mm