Learn More
Lenz Laborglasintrumente Three-Neck Round-Bottom Flasks Side Necks Parallel
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
867.08 SEK - 5326.50 SEK
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Standard Taper Joint | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Standard Taper Joint | Price | Quantity & Availability | |||
10498782
|
Lenz Laborglasintrumente
03304270 |
1 L | 131 mm | Center Neck NS 29/32, Side Necks NS 19/26 |
867.08 SEK
Each |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||
10473681
|
Lenz Laborglasintrumente
03304173 |
2 L | 166 mm | Center Neck NS 29/32, Side Necks NS 14/23 |
1309.10 SEK
Each |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||
10179550
|
Lenz Laborglasintrumente
03305482 |
5 L | 223 mm | Center Neck NS 45/40, Side Necks NS 29/32 |
3651.70 SEK
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
|||
10040321
|
Lenz Laborglasintrumente
03304488 |
10 L | 279 mm | Center Neck NS 29/32, Side Necks NS 29/32 |
5326.50 SEK
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
|||
10264431
|
Lenz Laborglasintrumente
03304476 |
3 L | 185 mm | Center Neck NS 29/32, Side Necks NS 29/32 |
2035.20 SEK
Each |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||
10274191
|
Lenz Laborglasintrumente
03304176 |
3 L | 185 mm | Center Neck NS 29/32, Side Necks NS 14/23 |
2167.70 SEK
Each |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||
10395921
|
Lenz Laborglasintrumente
03304485 |
6 L | 236 mm | Center Neck NS 29/32, Side Necks NS 29/32 |
3233.00 SEK
Each |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||
10438212
|
Lenz Laborglasintrumente
03304179 |
4 L | 207 mm | Center Neck NS 29/32, Side Necks NS 14/23 |
2443.30 SEK
Each |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||
10449172
|
Lenz Laborglasintrumente
03304482 |
5 L | 223 mm | Center Neck NS 29/32, Side Necks NS 29/32 |
3190.60 SEK
Each |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||
10738271
|
Lenz Laborglasintrumente
03304185 |
6 L | 236 mm | Center Neck NS 29/32, Side Necks NS 14/23 |
3105.80 SEK
Each |
Estimated Shipment: 15-11-2024
Log in to see stock. |
|||
Description
- Reaction flasks acc. to DIN 12392
- With two parallel side necks
- With conical ground joints
- With inscription label
- Material: DURAN™
Specifications
Duran Glass |