missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin(II) fluorophosphate, Thermo Scientific Chemicals
655.00 SEK - 2030.00 SEK
Chemical Identifiers
CAS | 52262-58-5 |
---|---|
Molecular Formula | FO3PSn |
Molecular Weight (g/mol) | 216.679 |
MDL Number | MFCD00058794 |
InChI Key | ZIQLCYCKKROENE-UHFFFAOYSA-L |
Synonym | tin ii fluorophosphate, stannous fluorophosphate, acmc-20ajhz, fluoridophosphoric acid tin ii salt, tin, ion sn2+ fluoro-phosphite ion, fluoro-dioxido-oxophosphorane; tin 2+, fluoranyl-bis oxidanidyl-oxidanylidene-$l^ 5-phosphane; tin 2+ |
PubChem CID | 44717639 |
IUPAC Name | fluoro-dioxido-oxo-$l^{5}-phosphane;tin(2+) |
SMILES | [O-]P(=O)([O-])F.[Sn+2] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52262-58-5 | |
216.679 | |
ZIQLCYCKKROENE-UHFFFAOYSA-L | |
44717639 | |
[O-]P(=O)([O-])F.[Sn+2] |
FO3PSn | |
MFCD00058794 | |
tin ii fluorophosphate, stannous fluorophosphate, acmc-20ajhz, fluoridophosphoric acid tin ii salt, tin, ion sn2+ fluoro-phosphite ion, fluoro-dioxido-oxophosphorane; tin 2+, fluoranyl-bis oxidanidyl-oxidanylidene-$l^ 5-phosphane; tin 2+ | |
fluoro-dioxido-oxo-$l^{5}-phosphane;tin(2+) |
Specifications
52262-58-5 | |
Powder | |
MFCD00058794 | |
Slightly soluble (3.0g/L at 25°C) | |
[O-]P(=O)([O-])F.[Sn+2] | |
216.679 | |
216.66 |
White | |
FO3PSn | |
tin ii fluorophosphate, stannous fluorophosphate, acmc-20ajhz, fluoridophosphoric acid tin ii salt, tin, ion sn2+ fluoro-phosphite ion, fluoro-dioxido-oxophosphorane; tin 2+, fluoranyl-bis oxidanidyl-oxidanylidene-$l^ 5-phosphane; tin 2+ | |
ZIQLCYCKKROENE-UHFFFAOYSA-L | |
fluoro-dioxido-oxo-$l^{5}-phosphane;tin(2+) | |
44717639 | |
Tin(II) fluorophosphate |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only