Learn More
Sonics Materials™ Titanium Alloy Half Wave Extender
Brand:  Sonics Materials™ 630-0409
Product Code. 11793129
  • 3955.00 SEK / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Specifications
Half Wave Extender
127 mm
1 Ea.
25 mm
Titanium Alloy
Documents
One moment while we fetch your results.
Product Title
Sonics Materials™ Titanium Alloy Half Wave Extender > Diameter: 25mm