missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TM 5275 sodium salt

Plasminogen activator inhibitor-1 (PAI-1) inhibitor; orally bioavailable

2195.00 SEK - 8505.00 SEK

Specifications

Chemical Name or Material 5-Chloro-2-[[2-[2-[4-(Diphenylmethyl)-1-piperazinyl]-2-oxoethoxy]acetyl]amino]benzoic acid sodium salt
CAS 1103926-82-4
Target Additional Synthetic Protease Inhibitors
Molecular Formula C28H27ClN3NaO5
Formula Weight 543.97
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18708219
View Documents
Tocris Bioscience™
5769/10
10 mg
2195.00 SEK
10mg
Please to purchase this item. Need a web account? Register with us today!
18719927
View Documents
Tocris Bioscience™
5769/50
50 mg
8505.00 SEK
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Plasminogen activator inhibitor-1 (PAI-1) inhibitor. Orally bioavailable.
Specifications

Specifications

5-Chloro-2-[[2-[2-[4-(Diphenylmethyl)-1-piperazinyl]-2-oxoethoxy]acetyl]amino]benzoic acid sodium salt
Additional Synthetic Protease Inhibitors
543.97
1103926-82-4
C28H27ClN3NaO5
>98%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.