missing translation for 'onlineSavingsMsg'
Learn More

TMEM190 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen TMEM190
Concentration 0.22 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16883415
View Documents Promotion Details
Proteintech
24904-1-AP-20UL
20 μL
1656.73 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16873415
View Documents Promotion Details
Proteintech
24904-1-AP-150UL
150 μL
4721.67 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TMEM190 gene ontology annotations related to this gene include hematopoietic progenitor cell differentiation.
Specifications

Specifications

TMEM190
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Q8WZ59
147744
TMEM190 Fusion Protein Ag19131
Primary
-20°C
TMEM190
0.22 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
MDAC1, TMEM190, transmembrane protein 190
TMEM190
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications