missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Tolvaptan
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 g |
Melting Point | 225.9°C |
Formula Weight | 448.95 |
InChI Key | GYHCTFXIZSNGJT-XMMPIXPASA-N |
SMILES | CC1=CC=CC=C1C(=O)NC1=CC=C(C(=O)N2CCC[C@@H](O)C3=CC(Cl)=CC=C23)C(C)=C1 |
IUPAC Name | N-{4-[(5R)-7-chloro-5-hydroxy-2,3,4,5-tetrahydro-1H-1-benzazepine-1-carbonyl]-3-methylphenyl}-2-methylbenzamide |
Molecular Weight (g/mol) | 448.95 |
Chemical Name or Material | Tolvaptan |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'