missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Torin 2

mTOR pathway inhibitor; Inhibits mTOR

1883.25 SEK - 3551.85 SEK

Specifications

CAS 1223001-51-1
Molecular Formula C24H15F3N4O
Purity ≥98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17139981
Encompass_Preferred
View Documents Promotion Details
STEMCELL Technologies
100-0259
5 mg
1883.25 SEK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17149981
Encompass_Preferred
View Documents Promotion Details
STEMCELL Technologies
100-0260
10 mg
3551.85 SEK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Torin 2 is a potent and selective ATP-competitive inhibitor against mTOR (EC50 = 0.25 nM), a key regulator of cell growth, survival, and autophagy (Liu et al.; Petherick et al.)
  • Inhibits proliferation of lung, breast, colorectal, and cervical cancer cell lines (Liu et al.)
  • Blocks mTOR complex 1 (mTORC1)-associated cell cycle progression and induces autophagy in hepatocellular carcinoma cells (Wang et al.)
Specifications

Specifications

1223001-51-1
C24H15F3N4O
≥98%
Videos
SDS
Documents

Documents

Product Certifications