Learn More
Hanna Instruments™ Total Chlorine Reagents
Prepared with high purity chemicals, high quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements.
Brand:  Hanna Instruments™ HI9371101
Product Code. 11897853
  • 779.00 SEK / Pack of 100
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Description
  • Reagents for the colorimetric determination of total chlorine
  • These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet for traceability
  • Pre-made reagents for ease of use
  • Prepared with high purity chemicals
  • High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements
Specifications
Specifications
Total Chlorine/0 to 350 mg