Learn More
Hanna Instruments™ Total Chlorine Reagents
Prepared with high purity chemicals, high quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements.
Brand: Hanna Instruments™ HI9371101
This item is not returnable.
View return policy
Description
- Reagents for the colorimetric determination of total chlorine
- These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet for traceability
- Pre-made reagents for ease of use
- Prepared with high purity chemicals
- High quality reagents that are pre-measured, allowing for users to achieve fast and accurate colorimetric measurements
Specifications
Total Chlorine/0 to 350 mg |