Learn More
Tocris Bioscience™ Toyocamycin
Adenosine analog; antifungal antibiotic
Brand:  Tocris Bioscience™ 5270/10
777.52 SEK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18764719
  • 885.00 SEK / 10mg
Explore more special offers
Description
Description
Toyocamycin is an adenosine analog; antifungal antibiotic. Inhibits RNA self-cleavage in HEK79 cells and PI 3-kinase activity in A431 epidermoid carcinoma cell membrane fractions. Also inhibits thapsigargin -induced XBP1-luciferase activation and induces apoptosis in multiple myeloma cell lines.
Specifications
Specifications
>98%
Toyocamycin
Toyocamycin
10 mg
291.26
Special Offers
For Research Use Only