missing translation for 'onlineSavingsMsg'
Learn More

TPI1 Monoclonal Antibody (P181), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA518291

Product Code. 15517251

  • 4230.00 SEK / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TPI1 Monoclonal antibody specifically detects TPI1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TPI1
Monoclonal
1 mg/mL
PBS with no preservative
P17751, P48500, P60174
Tpi1
Recombinant fragment corresponding to a region within amino acids 187 and 249 of Triosephosphate isomerase
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG3
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
P181
Unconjugated
Tpi1
AI255506; epididymis secretory protein Li 49; HEL-S-49; Methylglyoxal synthase; MGC88108; TIM; Tpi; TPI1; Tpi-1; TPID; triosephosphate isomerase; triose-phosphate isomerase; triosephosphate isomerase 1; triose-phosphate isomerase TPI1; unnamed protein product; YD9609.05C; YDR050C
Mouse
Protein G
RUO
21991, 24849, 7167
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.