missing translation for 'onlineSavingsMsg'
Learn More

TPT1/TCTP Rabbit anti-Human, Mouse, Rat, Clone: 9H7S4, Novus Biologicals™

Rabbit Monoclonal Antibody

1905.00 SEK - 4805.00 SEK

Specifications

Antigen TPT1/TCTP
Clone 9H7S4
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18326526
View Documents
Bio-Techne
NBP3-16122-20UL
20 μg
1905.00 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
18323515
View Documents
Bio-Techne
NBP3-16122-100UL
100 μg
4805.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TPT1/TCTP Monoclonal antibody specifically detects TPT1/TCTP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

TPT1/TCTP
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
Human, Mouse, Rat
FLJ27337, Fortilin, Histamine-releasing factor, p02, p23, TCTPHRFfortilin, translationally-controlled tumor protein, tumor protein, translationally-controlled 1
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TPT1/TCTP (P13693).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
9H7S4
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Stem Cell Markers
PBS, 0.05% BSA, 50% glycerol, pH7.3
7178
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.