Learn More
Fisherbrand™ Traceable™ Silicone Pump Tubing
Ideal for use with food, pharmaceuticals, and other critical solutions
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Description
Ideal for use with food, pharmaceuticals, and other critical solutions
- Replacement tubing for Variable-Speed Pump
- Can be sterilized by autoclaving
Specifications
Pump Tubing | |
1/16 in. | |
Silicone | |
7.6 m |
Autoclavable | |
1.58 mm | |
25 ft. | |
All Variable-Speed pumps |