missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF3IP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549934
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
The antibody detects endogenous levels of total TRAF3IP3 / T3JAM protein.
The gene encodes a protein that mediates cell growth by modulating the c-Jun N-terminal kinase signal transduction pathway. The encoded protein may also interact with a large multi-protein assembly containing the phosphatase 2A catalytic subunit. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Dec 2013].
Specifications
TRAF3IP3 | |
Polyclonal | |
Unconjugated | |
TRAF3IP3 | |
6030423D04Rik; AI115021; DJ434O14.3; FLJ44151; MGC117354; MGC163289; RGD1304848; T3JAM; TRAF3 interacting Jun N terminal kinase (JNK) activating modulator; TRAF3 interacting protein 3; TRAF3-interacting JNK-activating modulator; TRAF3-interacting JNK-activating modulator-like protein; TRAF3-interacting Jun N-terminal kinase (JNK)-activating modulator; TRAF3-interacting protein 3; Traf3ip3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
215243, 360900, 80342 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8C0G2, Q9Y228 | |
TRAF3IP3 | |
Synthesized peptide derived from internal of human T3JAM. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TRAF3IP3 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction