missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-trans-1,2-Diaminocyclohexane, 99%, Thermo Scientific Chemicals

Product Code. 11386926
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(±)-trans-1,2-Diaminocyclohexane is employed in the synthesis of macrocyclic [3+3] hexa Schiff base. It also plays an important role in the preparation of multidentate ligands, chiral auxiliaries and chiral stationary phases. Further, it is used to prepare [2+2] macrocyclization by reacting with aliphatic dialdehydes. In addition to this, it acts as a chiral ligand, which finds application in asymmetric catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(^+)-trans-1,2-Diaminocyclohexane is employed in the synthesis of macrocyclic [3+3] hexa Schiff base. It also plays an important role in the preparation of multidentate ligands, chiral auxiliaries and chiral stationary phases. Further, it is used to prepare [2+2] macrocyclization by reacting with aliphatic dialdehydes. In addition to this, it acts as a chiral ligand, which finds application in asymmetric catalysis.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-22-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00063747
InChI Key SSJXIUAHEKJCMH-PHDIDXHHSA-N
Synonym 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
PubChem CID 43806
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine
SMILES C1CCC(C(C1)N)N

Specifications

Melting Point 14°C to 15°C
Density 0.951
Boiling Point 89°C to 90°C (14 mmHg)
Flash Point 68°C (154°F)
Odor Amine-like
Refractive Index 1.489
Quantity 50 g
UN Number UN2735
Beilstein 3193807
Sensitivity Air sensitive; May form a precipitate
Solubility Information Miscible with water.
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material (+/-)-trans-1,2-Diaminocyclohexane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'