missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dibromocyclohexane, 99%, Thermo Scientific Chemicals

Product Code. 15450288
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

trans-1,2-Dibromocyclohexane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Dibromocyclohexane is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-37-0
Molecular Formula C6H10Br2
Molecular Weight (g/mol) 241.95
MDL Number MFCD00003820
InChI Key CZNHKZKWKJNOTE-PHDIDXHHSA-N
Synonym trans-1,2-dibromocyclohexane, 1r,2r-1,2-dibromocyclohexane, +/--trans-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, trans-1,2-dibromcyclohexan, 1beta,2alpha-dibromocyclohexane, cyclohexane,2-dibromo-, trans, 1r,2r-1,2-bis bromanyl cyclohexane
PubChem CID 98506
IUPAC Name (1R,2R)-1,2-dibromocyclohexane
SMILES Br[C@@H]1CCCC[C@H]1Br

Specifications

Density 1.784
Boiling Point 145°C (100 mmHg)
Flash Point >113°C (235°F)
Refractive Index 1.5515
Quantity 25 g
Formula Weight 241.95
Percent Purity 99%
Chemical Name or Material trans-1,2-Dibromocyclohexane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'