Learn More
trans-2-Ethoxyethenyl-1-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
1990.00 SEK - 5345.00 SEK
Chemical Identifiers
CAS | 1201905-61-4 |
---|---|
Molecular Formula | C10H19BO3 |
Molecular Weight (g/mol) | 198.07 |
MDL Number | MFCD09702441,MFCD09998813 |
InChI Key | MRAYNLYCQPAZJN-UHFFFAOYSA-N |
Synonym | e-2-2-ethoxyvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-2-ethoxyvinylboronic acid pinacol ester, 2-e-2-ethoxyethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-ethoxyvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-1-ethoxyethene-2-boronic acid pinacol ester, 2-2-ethoxyethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-1-ethoxyethene-2-ylboronic acid pinacol ester, z-2-ethoxyethenyl boronic acid, pinacol ester, trans-2-ethoxyethenyl-1-boronic acid pinacol esterShow More |
PubChem CID | 21973908 |
IUPAC Name | 2-[(E)-2-ethoxyethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | CCOC=CB1OC(C)(C)C(C)(C)O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458425
|
Thermo Scientific Alfa Aesar
H33367.03 |
1 g |
1990.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15468425
|
Thermo Scientific Alfa Aesar
H33367.06 |
5 g |
5345.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesMoisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
1201905-61-4 | |
198.07 | |
MRAYNLYCQPAZJN-UHFFFAOYSA-N | |
21973908 | |
CCOC=CB1OC(C)(C)C(C)(C)O1 |
C10H19BO3 | |
MFCD09702441,MFCD09998813 | |
e-2-2-ethoxyvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-2-ethoxyvinylboronic acid pinacol ester, 2-e-2-ethoxyethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-ethoxyvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-1-ethoxyethene-2-boronic acid pinacol ester, 2-2-ethoxyethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-1-ethoxyethene-2-ylboronic acid pinacol ester, z-2-ethoxyethenyl boronic acid, pinacol ester, trans-2-ethoxyethenyl-1-boronic acid pinacol esterShow More | |
2-[(E)-2-ethoxyethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
1201905-61-4 | |
C10H19BO3 | |
MFCD09702441,MFCD09998813 | |
MRAYNLYCQPAZJN-UHFFFAOYSA-N | |
2-[(E)-2-ethoxyethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
21973908 | |
98% |
50°C (0.3 mmHg) | |
1.446 | |
e-2-2-ethoxyvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-2-ethoxyvinylboronic acid pinacol ester, 2-e-2-ethoxyethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-ethoxyvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-1-ethoxyethene-2-boronic acid pinacol ester, 2-2-ethoxyethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, e-1-ethoxyethene-2-ylboronic acid pinacol ester, z-2-ethoxyethenyl boronic acid, pinacol ester, trans-2-ethoxyethenyl-1-boronic acid pinacol ester | |
CCOC=CB1OC(C)(C)C(C)(C)O1 | |
198.07 | |
198.07 | |
trans-2-Ethoxyethenyl-1-boronic acid pinacol ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.