missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexenoic acid, 96%, Thermo Scientific Chemicals

Product Code. 11430084
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13419-69-7
Molecular Formula C6H9O2
Molecular Weight (g/mol) 113.14
MDL Number MFCD00002705
InChI Key NIONDZDPPYHYKY-SNAWJCMRSA-M
Synonym trans-2-hexenoic acid, 2-hexenoic acid, e-hex-2-enoic acid, hex-2-enoic acid, trans-hex-2-enoic acid, e-2-hexenoic acid, hexenoic acid, 2e-hex-2-enoic acid, 2-hexenoic acid, 2e, 2-hexenoic acid, e
PubChem CID 5282707
ChEBI CHEBI:87721
IUPAC Name (E)-hex-2-enoic acid
SMILES CCC\C=C\C([O-])=O

Specifications

Melting Point 30°C to 35°C
Density 0.965
Boiling Point 217°C
Flash Point 125°C (257°F)
Refractive Index 1.4385
Quantity 10 g
UN Number UN3261
Beilstein 1720443
Formula Weight 114.14
Percent Purity 96%
Chemical Name or Material trans-2-Hexenoic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.