Learn More
trans-3-Aminocyclohexanecarboxamide hydrochloride, 97%, Thermo Scientific Chemicals
2560.00 SEK - 7555.00 SEK
Chemical Identifiers
CAS | 920966-29-6 |
---|---|
Molecular Formula | C7H15ClN2O |
Molecular Weight (g/mol) | 178.66 |
MDL Number | MFCD12022628 |
InChI Key | ZCFBGXVDLNNMDE-KGZKBUQUSA-N |
Synonym | trans-3-aminocyclohexanecarboxamide hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxamide hydrochloride |
PubChem CID | 46172883 |
IUPAC Name | (1R,3R)-3-aminocyclohexane-1-carboxamide;hydrochloride |
SMILES | C1CC(CC(C1)N)C(=O)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15447536
|
Thermo Scientific Alfa Aesar
H62529.MD |
250 mg |
2560.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15437536
|
Thermo Scientific Alfa Aesar
H62529.03 |
1 g |
7555.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
920966-29-6 | |
178.66 | |
ZCFBGXVDLNNMDE-KGZKBUQUSA-N | |
46172883 | |
C1CC(CC(C1)N)C(=O)N.Cl |
C7H15ClN2O | |
MFCD12022628 | |
trans-3-aminocyclohexanecarboxamide hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxamide hydrochloride | |
(1R,3R)-3-aminocyclohexane-1-carboxamide;hydrochloride |
Specifications
920966-29-6 | |
MFCD12022628 | |
ZCFBGXVDLNNMDE-KGZKBUQUSA-N | |
(1R,3R)-3-aminocyclohexane-1-carboxamide;hydrochloride | |
46172883 | |
97% |
C7H15ClN2O | |
trans-3-aminocyclohexanecarboxamide hydrochloride, 1r,3r-3-aminocyclohexane-1-carboxamide hydrochloride | |
C1CC(CC(C1)N)C(=O)N.Cl | |
178.66 | |
178.65 | |
trans-3-Aminocyclohexanecarboxamide hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.