missing translation for 'onlineSavingsMsg'
Learn More
trans-4-(4-n-Pentylcyclohexyl)phenyl trans,trans-4'-n-propylbicyclohexyl-4-carboxylate, 97%, Thermo Scientific™ Product Code.: 15405295

trans-4-(4-n-Pentylcyclohexyl)phenyl trans,trans-4'-n-propylbicyclohexyl-4-carboxylate, 97%, Thermo Scientific™

Product Code. 15405295
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15405295

Brand: Thermo Scientific Alfa Aesar H55324.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131790-57-3
Molecular Formula C33H52O2
Molecular Weight (g/mol) 480.78
MDL Number MFCD11053459,MFCD20527223
InChI Key HNSOCAVSNSWNJJ-UHFFFAOYSA-N
Synonym trans,trans-4-trans-4-pentylcyclohexyl-phenyl 4'-propylbicyclohexyl-4-carboxylate, 4-4-n-pentylcyclohexyl phenyl 4'-n-propylbicyclohexyl-4-carboxylate, 4-4-pentylcyclohexyl phenyl 4'-propyl-1,1'-bi cyclohexane-4-carboxylate, 4-4-pentylcyclohexyl phenyl 4'-propyl 1,1'-bi cyclohexane-4-carboxylate, trans,trans-4-trans-4-pentylcyclohexyl-phenyl 4-propylbicyclohexyl-4-carboxylate, trans-4-4-n-pentylcyclohexyl phenyl trans,trans-4'-n-propylbicyclohexyl-4-carboxylate, 1alpha-4beta-1alpha-4beta-propylcyclohexyl cyclohexanecarboxylic acid 4-1alpha-4beta-pentylcyclohexyl phenyl ester, trans,trans-4-4-trans-pentylcyclohexyl phenyl 4'-propyl-1,1'-bi cyclohexane-4-carboxylate, 4-4-pentylcyclohexyl phenyl 1r,1's,4r,4'r-4'-propyl-1,1'-bi cyclohexane-4-carboxylate, 4-1s,4r-4-pentylcyclohexyl phenyl 1r,1's,4r,4'r-4'-propyl-1,1'-bi cyclohexane-4-carboxylateShow MoreShow Less
PubChem CID 14993612
IUPAC Name [4-(4-pentylcyclohexyl)phenyl] 4-(4-propylcyclohexyl)cyclohexane-1-carboxylate
SMILES CCCCCC1CCC(CC1)C1=CC=C(OC(=O)C2CCC(CC2)C2CCC(CCC)CC2)C=C1

Specifications

Melting Point 213°C to 223°C
Quantity 5 g
Formula Weight 452.72
Percent Purity 97%
Chemical Name or Material trans-4-(4-n-Pentylcyclohexyl)phenyl trans,trans-4'-n-propylbicyclohexyl-4-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberBI090560 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-4-(4-n-Pentylcyclohexyl)phenyl trans,trans-4'-n-propylbicyclohexyl-4-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.