Learn More
Fisherbrand™ Translucent Silicone Tubing
Providing high tear strength tubing for the laboratory. Fisherbrand™ Translucent Silicone Tubing is available in a comprehensive range of sizes.
1490.00 SEK - 1780.00 SEK
Specifications
Certifications/Compliance | FDA, BFR and WRAS approved |
---|---|
Autoclavable | Autoclavable |
Material | Silicone |
Overall Length (Metric) | 4 m |
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
11746599
|
Fisherbrand™
|
9 mm |
1490.00 SEK
Each |
Estimated Shipment: 26-08-2024
Log in to see stock. |
|||||
11756599
|
Fisherbrand™
|
12 mm |
1780.00 SEK
Each |
Estimated Shipment: 25-09-2024
Log in to see stock. |
|||||
Description
- Autoclavable
- Non-yellowing
- High gloss finish
- Excellent quality
- Extended life in Peristaltic pumps
- Applications include biological, pharmaceutical, biotechnology, aerospace and high temperature work
Specifications
FDA, BFR and WRAS approved | |
Autoclavable | |
Silicone | |
4 m |