Learn More
VITLAB™ Transparent LDPE Sample Containers
Store liquid or solid samples securely. VITLAB™ Transparent LDPE Sample Containers feature an attached snap-on lid to protect samples from contamination, leaking or spillage. Transparent bottles ensure easy identification. Available in a variety of sizes.
Brand:  VITLAB™ 80735
 View more versions of this product
Product Code. 10166210
Description
Description
Economical, ready-to-use containers ideal for storing a wide variety of liquid and solid materials.
  • Constructed of durable, break-resistant low-density polyethylene (LDPE)
  • Transparent bottles allow for easy identification of contents
  • Snap-on lids prevent spills and leaks, even when the bottle is upside down
  • Attached lids keep closure accessible for one-handed use and can’t be lost
Specifications
Specifications
50 mL
Storing a Wide Variety of Liquid and Solid Materials
LDPE
0.1 kg
Round
Sample Container
30 mm
Transparent
Plug Seal Cap
50Pack
Transparent
74 mm
LDPE
Plug Seal