missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAX Rabbit anti-Human, Mouse, Rat, Clone: S02-3D3, Novus Biologicals™
Rabbit Monoclonal Antibody
2825.00 SEK - 5705.00 SEK
Specifications
Antigen | TRAX |
---|---|
Clone | S02-3D3 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18353874
|
Bio-Techne
NBP3-15102-25UL |
25 μg |
2825.00 SEK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18322794
|
Bio-Techne
NBP3-15102-100UL |
100 μg |
5705.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRAX Monoclonal antibody specifically detects TRAX in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
TRAX | |
Western Blot | |
Unconjugated | |
Rabbit | |
Direct Reversal of DNA Damage | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
7257 | |
IgG | |
Affinity purified |
S02-3D3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DISC1, translin-associated factor Xdisrupted in schizophrenia 1, translin-like protein, TRAXtranslin-associated protein X | |
A synthetic peptide of human TRAX (Uniprot#: Q99598) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title