Learn More
Tri-n-butyltin isocyanate, 97%, Thermo Scientific™
233.00 SEK - 5058.00 SEK
Chemical Identifiers
CAS | 681-99-2 |
---|---|
Molecular Formula | C13H27NOSn |
Molecular Weight (g/mol) | 332.075 |
MDL Number | MFCD08272321 |
InChI Key | YGUNDMHXMXDGII-UHFFFAOYSA-N |
Synonym | tributyltin isocyanate, tributyl isocyanato stannane, stannane, isocyanato tributyl, tri-n-butyltin isocyanate, tributylstannyl isocyanate, tributyltin isothiocyanate, stannane, tributylisocyanato, tin, tributyl-, isothiocyanate, tri-n-butylisocyanatotin, acmc-20ajf1 |
PubChem CID | 120323 |
IUPAC Name | tributyl(isocyanato)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)N=C=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
681-99-2 | |
332.075 | |
YGUNDMHXMXDGII-UHFFFAOYSA-N | |
120323 | |
CCCC[Sn](CCCC)(CCCC)N=C=O |
C13H27NOSn | |
MFCD08272321 | |
tributyltin isocyanate, tributyl isocyanato stannane, stannane, isocyanato tributyl, tri-n-butyltin isocyanate, tributylstannyl isocyanate, tributyltin isothiocyanate, stannane, tributylisocyanato, tin, tributyl-, isothiocyanate, tri-n-butylisocyanatotin, acmc-20ajf1 | |
tributyl(isocyanato)stannane |
Specifications
681-99-2 | |
1.203 g/mL | |
110°C (230°F) | |
[CH3(CH2)3]3SnNCO | |
MFCD08272321 | |
tributyltin isocyanate, tributyl isocyanato stannane, stannane, isocyanato tributyl, tri-n-butyltin isocyanate, tributylstannyl isocyanate, tributyltin isothiocyanate, stannane, tributylisocyanato, tin, tributyl-, isothiocyanate, tri-n-butylisocyanatotin, acmc-20ajf1 | |
CCCC[Sn](CCCC)(CCCC)N=C=O | |
332.075 | |
332.05 | |
Liquid |
Colorless | |
297°C | |
C13H27NOSn | |
1.489 | |
UN2788 | |
YGUNDMHXMXDGII-UHFFFAOYSA-N | |
tributyl(isocyanato)stannane | |
120323 | |
97% | |
Tri-n-butyltin isocyanate |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
missing translation for 'rtecsNumber' : WH8583500
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.