missing translation for 'onlineSavingsMsg'
Learn More
Tributylamine, ≥98.5%, Honeywell™ Product Code.: 15625630

Tributylamine, ≥98.5%, Honeywell™

Product Code. 15625630
250 mL, Glass Bottle
Click to view available options
Quantity:
25 mL
250 mL
2 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15625630

Brand: Honeywell Research Chemicals 471313250ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥98.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-82-9
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.355
MDL Number MFCD00009431
InChI Key IMFACGCPASFAPR-UHFFFAOYSA-N
Synonym tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7Show MoreShow Less
PubChem CID 7622
ChEBI CHEBI:38905
IUPAC Name N,N-dibutylbutan-1-amine
SMILES CCCCN(CCCC)CCCC

Specifications

Melting Point -70°C
Color Colorless
Density 0.778g/mL at 25°C
Boiling Point 216°C
Flash Point 63°C (145.4°F)
Odor Unpleasant
Packaging Glass Bottle
Linear Formula [CH3(CH2)3]3N
Quantity 250 mL
UN Number UN2542
Beilstein 1698872
Formula Weight 185.35g/mol
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Tributylamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tributylamine, ≥98.5%, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.