missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tributylamine, ≥98.5%, Honeywell™
Click to view available options
Quantity:
25 mL
250 mL
2 L
2.5 L
Packaging:
Glass Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Specifications
Specifications
Melting Point | -70°C |
Color | Colorless |
Density | 0.778g/mL at 25°C |
Boiling Point | 216°C |
Flash Point | 63°C (145.4°F) |
Odor | Unpleasant |
Packaging | Glass Bottle |
Linear Formula | [CH3(CH2)3]3N |
Quantity | 2 L |
UN Number | UN2542 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tributylamine, ≥98.5%, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction