missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic Acid 0.60 N Solution MP Biomedicals
Click to view available options
Quantity:
25 mL
Specifications
Molecular Formula | C2HCl3O2 |
Synonym | trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer |
InChI Key | YNJBWRMUSHSURL-UHFFFAOYSA-N |
SMILES | C(=O)(C(Cl)(Cl)Cl)O |
IUPAC Name | 2,2,2-trichloroacetic acid |
Molecular Weight (g/mol) | 163.378 |
PubChem CID | 6421 |
ChEBI | CHEBI:30956 |
Formula Weight | 163.4 |
Density | 5.6 (air = 1) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'