missing translation for 'onlineSavingsMsg'
Learn More

Trioctyl phosphate, Thermo Scientific Chemicals

312.00 SEK - 1845.00 SEK

Chemical Identifiers

CAS 78-42-2
Molecular Formula C24H51O4P
Molecular Weight (g/mol) 434.64
MDL Number MFCD00009491
InChI Key GTVWRXDRKAHEAD-UHFFFAOYNA-N
Synonym tris 2-ethylhexyl phosphate, disflamoll tof, flexol tof, phosphoric acid, tris 2-ethylhexyl ester, kronitex tof, flexol plasticizer tof, 2-ethyl-1-hexanol phosphate, tris ethylhexyl phosphate, tri 2-ethylhexyl phosphate
PubChem CID 6537
IUPAC Name tris(2-ethylhexyl) phosphate
SMILES CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307148
SDS View Documents
Thermo Scientific Alfa Aesar
015808.18
50 g
312.00 SEK
50g
Please to purchase this item. Need a web account? Register with us today!
11387138
SDS View Documents
Thermo Scientific Alfa Aesar
015808.30
250 g
734.00 SEK
250g
Please to purchase this item. Need a web account? Register with us today!
11397138
SDS View Documents
Thermo Scientific Alfa Aesar
015808.A1
1 kg
1845.00 SEK
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Trioctyl phosphate finds application as a fire-retardant, plasticizer for polyvinyl chloride and cellulose nitrate. It is used as a stimulant for the chemical warfare nerve agent VX. Further, it used as an alkylation agent for nitrogen heterocyclic compounds and as a catalyst to produce phenolic and urea resins. In addition to this, it is used as an antifoaming agent and a co-solvent in hydrogen peroxide production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78-42-2
434.64
GTVWRXDRKAHEAD-UHFFFAOYNA-N
6537
CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC
C24H51O4P
MFCD00009491
tris 2-ethylhexyl phosphate, disflamoll tof, flexol tof, phosphoric acid, tris 2-ethylhexyl ester, kronitex tof, flexol plasticizer tof, 2-ethyl-1-hexanol phosphate, tris ethylhexyl phosphate, tri 2-ethylhexyl phosphate
tris(2-ethylhexyl) phosphate

Specifications

78-42-2
216°C (4 mmHg)
C24H51O4P
MFCD00009491
Miscible with mineral oil and gasoline. Immiscible with water.
CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC
434.64
434.64
Trioctyl phosphate
0.93 g/mL
>110°C (230°F)
1.441
tris 2-ethylhexyl phosphate, disflamoll tof, flexol tof, phosphoric acid, tris 2-ethylhexyl ester, kronitex tof, flexol plasticizer tof, 2-ethyl-1-hexanol phosphate, tris ethylhexyl phosphate, tri 2-ethylhexyl phosphate
GTVWRXDRKAHEAD-UHFFFAOYNA-N
tris(2-ethylhexyl) phosphate
6537
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H332-H315-H319
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Call a POISON CENTER or doctor/physician if you feel unwell.

Warning

missing translation for 'einecsNumber' : 201-116-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.