Learn More
Triphenylphosphine dibromide, ca. 33% bromine, Thermo Scientific Chemicals
426.56 SEK - 3832.18 SEK
Chemical Identifiers
CAS | 1034-39-5 |
---|---|
Molecular Formula | C18H15Br2P |
Molecular Weight (g/mol) | 422.10 |
MDL Number | MFCD00000054 |
InChI Key | OCXGTPDKNBIOTF-UHFFFAOYSA-N |
Synonym | dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane |
PubChem CID | 70579 |
IUPAC Name | dibromo(triphenyl)-$l^{5}-phosphane |
SMILES | BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10787583
|
Thermo Scientific Acros
291070050 |
5 g | Plastic bottle |
426.56 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10134830
|
Thermo Scientific Acros
291070250 |
25 g | Plastic bottle |
1265.99 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10548883
|
Thermo Scientific Acros
291071000 |
100 g | Plastic bottle |
3832.18 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
1034-39-5 | |
422.10 | |
OCXGTPDKNBIOTF-UHFFFAOYSA-N | |
70579 | |
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C18H15Br2P | |
MFCD00000054 | |
dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane | |
dibromo(triphenyl)-$l^{5}-phosphane |
Specifications
1034-39-5 | |
Pink to Yellow | |
C18H15Br2P | |
MFCD00000054 | |
01,1247; 13,333 | |
dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane | |
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
422.10 | |
422.09 | |
Crystalline Powder |
235.0°C | |
Authentic | |
(C6H5)3PBr2 | |
16, 864 | |
15, 9917 | |
OCXGTPDKNBIOTF-UHFFFAOYSA-N | |
dibromo(triphenyl)-$l^{5}-phosphane | |
70579 | |
ca. 33% bromine | |
Triphenylphosphine dibromide, ca. 33% bromine |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-855-1
- Triphenylphosphine dibromide, ca. 33% bromine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only