missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine
Carry out environmental monitoring in the most regulated environments with Thermo Scientific™ Triple Wrapped Irradiated plates for air sampling applications.
3360.00 SEK
Specifications
Color | Ivory, transparent |
---|---|
Content And Storage | 2 to 25°C |
Description | Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine |
Manufacturing Origin | Germany |
Product Type | Agar |
Product Code | Brand | Description | Format | Quantity | Manufacturing Origin | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Format | Quantity | Manufacturing Origin | Price | Quantity & Availability | ||
15882467
|
Thermo Scientific™
PO5513D |
Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine | 55mm Contact Plate | 10/Pk. | Germany |
3360.00 SEK
Pack of 10 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||
Description
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Sabouraud Dextrose Agar plates, complete with VHP Indicator, Lecithin, Polysorbate 80, Sodium Thiosulphate and L-Histidine.
Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.Three protective layers reduce risk of contamination:
- Primary film with moisture control patch
- Secondary film VHP barrier with immobilized desiccant sachet
- Tertiary film dust cover
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.
Specifications
Ivory, transparent | |
Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine | |
Agar |
2 to 25°C | |
Germany | |
Triple Wrapped - Irradiated |
Safety and Handling
missing translation for 'shelfLife' : 12 Weeks