missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-thienyl)phosphine, 98+%, Thermo Scientific Chemicals

Product Code. 11405373
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It employed as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical and organic synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool, dry place in tightly closed container. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24171-89-9
Molecular Formula C12H9PS3
Molecular Weight (g/mol) 280.36
MDL Number MFCD00015471
InChI Key KUCPTMZJPDVWJL-UHFFFAOYSA-N
Synonym tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine #
PubChem CID 90384
IUPAC Name trithiophen-2-ylphosphane
SMILES S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1

Specifications

Melting Point 28°C to 30°C
Odor Recognizable
Quantity 25 g
Beilstein 203736
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 280.37
Percent Purity ≥98%
Chemical Name or Material Tris(2-thienyl)phosphine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'