missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride, MP Biomedicals™
Click to view available options
Quantity:
100 g
250 g
500 g
Description
Uses include:
- pH control in vitro and in vivo for body fluids
- In buffering systems for electrophoresis applications
- As a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes)
- Direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid
Specifications
Physical Form | Crystalline Powder |
Quantity | 250 g |
Solubility Information | Soluble in water (550mg/mL), ethylene glycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL) |
Formula Weight | 157.6 |
Percent Purity | >99% |
Grade | Reagent |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'