Learn More
Tropisetron Hydrochloride, MedChemExpress
MedChemExpress Tropisetron Hydrochloride (SDZ-ICS-930) is a selective 5-HT3 receptor antagonist and α7-nicotinic receptor agonist with an IC50 of 70.1 ± 0.9 nM for 5-HT3 receptor.
Brand: Medchem Express HY-B0020
Description
Tropisetron Hydrochloride (SDZ-ICS-930) is a selective 5-HT3 receptor antagonist and α7-nicotinic receptor agonist with an IC50 of 70.1 ± 0.9 nM for 5-HT3 receptor.Specifications
Tropisetron Hydrochloride | |
105826-92-4 | |
10 mg | |
C17H21ClN2O2 | |
SDZ-ICS-930 | |
O=C(C1=CNC2=C1C=CC=C2)O[C@H]3C[C@H]4CC[C@H](N4C)C3.[H]Cl | |
320.81 | |
Research |
Solid | |
White | |
Research | |
Neuroscience-Neuromodulation | |
H2O : ≥ 50 mg/mL (155.86 mM) ∣DMSO : 33.33 mg/mL (103.89 mM; Need ultrasonic) | |
320.81 | |
99.95% |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only