missing translation for 'onlineSavingsMsg'
Learn More

Trypsin inhibitor, MP Biomedicals™

Product Code. 11368699
Click to view available options
Quantity:
25 mg
100 mg
250 mg
500 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 9035-81-8
Synonym Ovomucoid trypsin inhibitor

Specifications

Color White
Quantity 25 mg
Formula Weight 28,500 ± 3,5003,4,5
Physical Form Powder
Chemical Name or Material Trypsin Inhibitor
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.