missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Tryptone Water (Peptone Water)
Tryptone Water (Peptone Water) is used for the detection of indole production in coliform microorganisms.
Brand: Scharlab 03-156-500
This item is not returnable.
View return policy
Description
- COMPF/ISO/SMWW
- Substrate with low nutrient capacity, for the detection of indole production in coliform microorganisms
- According to ISO 7251 standard.
Specifications
ISO 7251 | |
Tryptone Water (Peptone Water) | |
Coliforms | |
Detection of Indol Production in Coliform Microorganisms | |
48 ± 2 hr. | |
500 g |
Solid | |
Agar | |
Casein Peptone 10.0 g/L, Sodium Chloride 5.0 g/L | |
44°C ± 0.5°C | |
7.3 ±0.2 | |
Insoluble |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Scharlab Tryptone Water (Peptone Water) > Tryptone Water; Form: Solid; Target Organism Class: Coliforms
Spot an opportunity for improvement?Share a Content Correction