missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Tube Racks, Short

Tube Racks, Short are designed for use on Multi Vortex-Genie, Mini Orbital-Genie 300 Ratcheting Clamp Models or Genie Temp-Shaker Ratcheting Clamp Models.

1680.00 SEK - 2705.00 SEK

Specifications

Holds Tubes
Tube Size Up to 29 mm dia.
Product Type Tube Rack
Color Clear
For Use With (Application) Shaking of Various Size and Material Tubes
View More Specs

Products 2
Product Code Brand Packaging Quantity Price Quantity & Availability  
Product Code Brand Packaging Quantity Price Quantity & Availability  
17231762
View Documents Promotion Details
Scientific Industries
SI-M010S
1 Ea.
1680.00 SEK
Each
Estimated Shipment: 20-11-2024
to see stock.
Add to basket
 
17241762
View Documents Promotion Details
Scientific Industries
SI-M020S
2/Pk.
2705.00 SEK
Pack of 2
Estimated Shipment: 20-11-2024
to see stock.
Add to basket
 
Description

Description

  • Accommodates tubes of any material
Specifications

Specifications

Tubes
Tube Rack
Shaking of Various Size and Material Tubes
12
Up to 29 mm dia.
Clear
Multi Vortex-Genie, Mini Orbital-Genie 300 Ratcheting Clamp Models/Genie Temp-Shaker Ratcheting Clamp Models
Videos
SDS
Documents

Documents

Product Certifications