missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SymLine FlexTube Screw Connection
Tubing screw connection constructed of chemically resistant PTFE.
1030.00 SEK - 1070.00 SEK
Specifications
Product Type | Screw Connection |
---|---|
Material | PTFE |
Packaging Quantity | 2 Pk. |
Color | Black |
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
15840945
|
SymLine
106579 |
FlexTube GL25 Tubing |
1030.00 SEK
Pack of 2 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
15813652
|
SymLine
106591 |
FlexTube Pro GL25 Tubing |
1070.00 SEK
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
|||||
Description
- PTFE construction for chemical resistance
- Secure, screw-type connection
- Suitable for FlexTube tubing
Specifications
Screw Connection | |
PTFE | |
2 Pk. | |
Black |